Learn More
Med Vet International Dynarex Full Length Face Shield, Each
Catalog No. 502092442
Encompass_Preferred
Full length face shield with a wrap-around design provides the user with peripheral vision Protects against non-hazardous liquid splash and particles Fits easily over prescription glasses or protective eyewear Anti-fog coating reduces fogging in a high humidity environment. Disposable, one time use Elastic headband with foam for comfort
Catalog No. 50-209-2442 Supplier Med Vet International Supplier No. MASKFS
Full length face shield with a wrap-around design provides the user with peripheral vision Protects against non-hazardous liquid splash and particles Fits easily over prescription glasses or protective eyewear Anti-fog coating reduces fogging in a high humidity environment. Disposable, one time use Elastic headband with foam for comfort