Learn More
MSC Tape Logic Masking Tape, Black, 4.9 mil, 1 ea
Catalog No. 501918405
Encompass
Tape Logic Colored Masking Tape is suitable for medium-duty masking applications, arts & crafts, and hobbies. It can be used for color-coding, inventory identification, and medical coding. This tape seals securely across varied temperatures and humidity, conforming well to irregular surfaces.
  • Seals and sticks securely under a wide range of temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb per inch for break resistance
  • Easy to tear by hand
  • Users can write directly on the tape
Catalog No. 50-191-8405 Supplier MSC Supplier No. 39579958
Tape Logic Colored Masking Tape is suitable for medium-duty masking applications, arts & crafts, and hobbies. It can be used for color-coding, inventory identification, and medical coding. This tape seals securely across varied temperatures and humidity, conforming well to irregular surfaces.
  • Seals and sticks securely under a wide range of temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb per inch for break resistance
  • Easy to tear by hand
  • Users can write directly on the tape