Learn More
MSC Tape Logic Masking Tape, 1 in, Green, 1 ea
Catalog No. 501918389
Encompass
This green masking tape is designed for medium-duty masking applications and is suitable for arts & crafts and hobbies. It seals and sticks securely across various temperature and humidity conditions, conforming well to curves and irregular surfaces.
  • Made from crepe paper with a rubber adhesive
  • Tensile strength of 18 lb. per inch
  • Resistant to breaking and easy to tear by hand
  • Can be written on directly
  • Suitable for color coding shipments, identifying inventory, or medical coding applications
Catalog No. 50-191-8389 Supplier MSC Supplier No. 39579792
This green masking tape is designed for medium-duty masking applications and is suitable for arts & crafts and hobbies. It seals and sticks securely across various temperature and humidity conditions, conforming well to curves and irregular surfaces.
  • Made from crepe paper with a rubber adhesive
  • Tensile strength of 18 lb. per inch
  • Resistant to breaking and easy to tear by hand
  • Can be written on directly
  • Suitable for color coding shipments, identifying inventory, or medical coding applications