Learn More
MSC Tape Logic Masking Tape, 2 in. x 60 yd, 24/Pk
Catalog No. 501918407
Encompass
This masking tape is made from crepe paper with a rubber adhesive. It is 2 in. wide, 60 yd long, and 6.1 mil thick. The color of the tape is tan. It is suitable for heavy-duty masking, engineered for painting and construction applications.
  • Resistant to moisture and solvents
  • Seals and sticks securely under a wide range of temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb/in for break resistance
  • Easy to tear by hand
  • Allows for writing directly on the tape
Catalog No. 50-191-8407 Supplier MSC Supplier No. 39579974
This masking tape is made from crepe paper with a rubber adhesive. It is 2 in. wide, 60 yd long, and 6.1 mil thick. The color of the tape is tan. It is suitable for heavy-duty masking, engineered for painting and construction applications.
  • Resistant to moisture and solvents
  • Seals and sticks securely under a wide range of temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb/in for break resistance
  • Easy to tear by hand
  • Allows for writing directly on the tape