Learn More
Grainger Masking Tape Tan
Catalog No. 502767591
Encompass_Preferred
The 3M General Purpose Masking Tape is a versatile masking tape ideal for holding, bundling, sealing, splicing, and other general purpose applications. It features a crepe paper backing and rubber-based adhesive that provides a quick stick and clean removal. This general use masking tape is available in multiple widths from 1/4 inch to 2 inches and in standard 60 yard rolls. The 3M tape offers good adhesion and holding power while not leaving residue upon removal. It can be used for projects, crafts, home, and office use.
Catalog No. 50-276-7591 Supplier Grainger Supplier No. 16V085
The 3M General Purpose Masking Tape is a versatile masking tape ideal for holding, bundling, sealing, splicing, and other general purpose applications. It features a crepe paper backing and rubber-based adhesive that provides a quick stick and clean removal. This general use masking tape is available in multiple widths from 1/4 inch to 2 inches and in standard 60 yard rolls. The 3M tape offers good adhesion and holding power while not leaving residue upon removal. It can be used for projects, crafts, home, and office use.