Learn More
Grainger Masking Tape Tan PK100
Catalog No. 502766926
Encompass_Preferred
The 3M 1-inch masking tape is a general purpose masking tape that features a crepe paper backing and rubber-based adhesive. It is designed for holding, bundling, sealing, splicing, and masking applications. This tape has a temperature resistance up to 200°F and can be used on multiple surfaces including wood, glass, metal, and plastic. The 1-inch width allows for versatility in projects and applications. Overall, this is a reliable masking tape from 3M that provides good adhesion and clean removal.
Catalog No. 50-276-6926 Supplier Grainger Supplier No. 16V082
The 3M 1-inch masking tape is a general purpose masking tape that features a crepe paper backing and rubber-based adhesive. It is designed for holding, bundling, sealing, splicing, and masking applications. This tape has a temperature resistance up to 200°F and can be used on multiple surfaces including wood, glass, metal, and plastic. The 1-inch width allows for versatility in projects and applications. Overall, this is a reliable masking tape from 3M that provides good adhesion and clean removal.