Learn More
Grainger Masking Tape Tan PK100
Catalog No. 502767592
Encompass_Preferred
The 3M Painter's Tape is a multi-surface painter's tape that can be used on walls, woodwork, glass, and metal. It has a high tack adhesive that sticks well but removes cleanly without damage. The tape has a smooth surface and clean edges that resist paint bleed. It is available in several widths including 1", 1-1/2", and 2" and in lengths of 55 yards. The tape is designed for indoor and outdoor use and can withstand temperatures up to 200°F for 30 minutes. It leaves no sticky residue when removed within 14 days. This painter's tape delivers sharp paint lines and can be used for masking, bundling, and labeling.
Catalog No. 50-276-7592 Supplier Grainger Supplier No. 16V091
The 3M Painter's Tape is a multi-surface painter's tape that can be used on walls, woodwork, glass, and metal. It has a high tack adhesive that sticks well but removes cleanly without damage. The tape has a smooth surface and clean edges that resist paint bleed. It is available in several widths including 1", 1-1/2", and 2" and in lengths of 55 yards. The tape is designed for indoor and outdoor use and can withstand temperatures up to 200°F for 30 minutes. It leaves no sticky residue when removed within 14 days. This painter's tape delivers sharp paint lines and can be used for masking, bundling, and labeling.