Learn More
Grainger Masking Tape Tan PK125
Catalog No. 502767149
Encompass_Preferred
The Grainger 16V089 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing, sleeve bearing electric motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard NEMA mounting. This motor can provide 0.37 kW of output power.
Catalog No. 50-276-7149 Supplier Grainger Supplier No. 16V089
The Grainger 16V089 is a 1/2 HP, 115V, 1725 RPM, 56C frame, open drip proof, single phase, ball bearing, sleeve bearing electric motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with standard NEMA mounting. This motor can provide 0.37 kW of output power.