Learn More
Grainger Masking Tape Tan PK2500
Catalog No. 502767631
Encompass_Preferred
The 3M Multi-Use General Purpose Masking Tape is a 60 yard roll of 1.88 inch wide tan colored crepe paper masking tape. It features a rubber-based adhesive that provides quick stick and clean removal up to 14 days after application. The tape is designed for indoor masking, bundling, sealing, splicing, and other general purpose applications. It can be used on multiple surfaces including wood, glass, metal, and plastic. The tape has a temperature resistance range of up to 200°F.
Catalog No. 50-276-7631 Supplier Grainger Supplier No. 16V080
The 3M Multi-Use General Purpose Masking Tape is a 60 yard roll of 1.88 inch wide tan colored crepe paper masking tape. It features a rubber-based adhesive that provides quick stick and clean removal up to 14 days after application. The tape is designed for indoor masking, bundling, sealing, splicing, and other general purpose applications. It can be used on multiple surfaces including wood, glass, metal, and plastic. The tape has a temperature resistance range of up to 200°F.