Learn More
Grainger Masking Tape Tan PK2500
Catalog No. 502767154
Encompass_Preferred
The 3M Multi-Use General Purpose Masking Tape is a 60 yard roll of 1.88 inch wide masking tape. It features a crepe paper backing and rubber-based adhesive that provides good holding power while removing cleanly with little residue. This tape is designed for indoor masking, bundling, sealing, and other general purpose applications. It can be used on multiple surfaces including wood, glass, metal, and plastic. The tape has a temperature resistance up to 200°F. This 3M masking tape comes in tan color and provides good value for general masking jobs.
Catalog No. 50-276-7154 Supplier Grainger Supplier No. 16V079
The 3M Multi-Use General Purpose Masking Tape is a 60 yard roll of 1.88 inch wide masking tape. It features a crepe paper backing and rubber-based adhesive that provides good holding power while removing cleanly with little residue. This tape is designed for indoor masking, bundling, sealing, and other general purpose applications. It can be used on multiple surfaces including wood, glass, metal, and plastic. The tape has a temperature resistance up to 200°F. This 3M masking tape comes in tan color and provides good value for general masking jobs.