Learn More
Grainger Masking Tape Tan PK2500
Catalog No. 502765969
Encompass_Preferred
The 3M 1 in. x 60 yd. General Use Masking Tape is a multi-purpose masking tape that can be used for holding, sealing, splicing, and labeling. It features a crepe paper backing and rubber-based adhesive. This tape is designed for indoor and short-term outdoor applications. It can adhere to metal, glass, rubber, plastic, and painted wood surfaces. The tape has a temperature resistance up to 200°F. It comes in a standard 1 in. width and 60 yd. length roll. The tape color is tan. This is a general use masking tape from 3M ideal for common masking, holding, sealing, and labeling applications.
Catalog No. 50-276-5969 Supplier Grainger Supplier No. 16V078
The 3M 1 in. x 60 yd. General Use Masking Tape is a multi-purpose masking tape that can be used for holding, sealing, splicing, and labeling. It features a crepe paper backing and rubber-based adhesive. This tape is designed for indoor and short-term outdoor applications. It can adhere to metal, glass, rubber, plastic, and painted wood surfaces. The tape has a temperature resistance up to 200°F. It comes in a standard 1 in. width and 60 yd. length roll. The tape color is tan. This is a general use masking tape from 3M ideal for common masking, holding, sealing, and labeling applications.