Learn More
MSC 3M 7000124879 12mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200�F Max, Series 201+
Catalog No. 501919909
Encompass
3M 7000124879 12mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200�F Max, Series 201+
This item is not returnable. View return policy
Catalog No. 50-191-9909 Supplier MSC Supplier No. 52649324
3M 7000124879 12mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200�F Max, Series 201+