Learn More
MSC 3M 7000144752 48mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200°F Max, Series 201+
Catalog No. 501919911
Encompass
3M 7000144752 48mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200°F Max, Series 201+
Catalog No. 50-191-9911 Supplier MSC Supplier No. 52649407
3M 7000144752 48mm Wide x 180' Long x 4.4 mil Thick Tan Masking Tape Paper Backing, Rubber Adhesive, 19 Lb/In Tensile Strength, 200°F Max, Series 201+