Learn More
MSC Tape Logic Masking Tape, Crepe Paper, 60 yd, 1 Roll
Catalog No. 501919404
Encompass
This masking tape is designed for medium-duty applications, featuring a crepe paper backing and rubber adhesive. It is suitable for various tasks including paint masking, splicing, bundling, and general-purpose use in diverse environments.
  • Designed for medium-duty masking applications
  • Suitable for arts and crafts and hobbies
  • Seals and sticks securely under various temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb. per inch for break resistance
  • Easy to tear by hand
  • Allows direct writing on the tape
  • Features improved strength for industrial use
  • Moisture and oil resistant backing and adhesive
Catalog No. 50-191-9404 Supplier MSC Supplier No. 39590179
This masking tape is designed for medium-duty applications, featuring a crepe paper backing and rubber adhesive. It is suitable for various tasks including paint masking, splicing, bundling, and general-purpose use in diverse environments.
  • Designed for medium-duty masking applications
  • Suitable for arts and crafts and hobbies
  • Seals and sticks securely under various temperature and humidity conditions
  • Conforms well around curves and irregular surfaces
  • Tensile strength of 18 lb. per inch for break resistance
  • Easy to tear by hand
  • Allows direct writing on the tape
  • Features improved strength for industrial use
  • Moisture and oil resistant backing and adhesive