Learn More
Medchemexpress LLC HY-N0629 5mg Medchemexpress, Maslinic acid CAS:4373-41-5 Purity:>98%
SDP
Catalog No. 501933648
Encompass_Preferred
Medchemexpress, HY-N0629 5mg Maslinic acid CAS:4373-41-5 Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α , which is required for p65 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3648 Supplier Medchemexpress LLC Supplier No. HYN06295MG
Medchemexpress, HY-N0629 5mg Maslinic acid CAS:4373-41-5 Maslinic acid can inhibit the DNA-binding activity of NF-κB p65 and abolish the phosphorylation of IκB-α , which is required for p65 activation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.