Learn More
Cayman Chemical Maslinic Acid Inhibitors
SDP
Catalog No. 502887505
Encompass_Preferred
An antiproliferative agent against Caco-2 cancer cells (EC50 = 15 µM), HT-29 human colon cancer cells (EC50 = 74 µM), 1321N1 astrocytoma cells (IC50 = 25 µM), and human leukemia (CCRF-CEM and CEM/ADR5000) cells (IC50 = 7 and 9 µM respectively)
Catalog No. 50-288-7505 Supplier Cayman Chemical Supplier No. 1000964510 MG
Add to Cart
Add to Cart
An antiproliferative agent against Caco-2 cancer cells (EC50 = 15 µM), HT-29 human colon cancer cells (EC50 = 74 µM), 1321N1 astrocytoma cells (IC50 = 25 µM), and human leukemia (CCRF-CEM and CEM/ADR5000) cells (IC50 = 7 and 9 µM respectively)