Learn More
Grainger Masonry Saw Dry 14 in Blade Dia
Catalog No. 502750907
Encompass_Preferred
The Makita 14-Inch Masonry Saw has a powerful 15.0 AMP motor that delivers 5,800 RPM for fast, efficient cutting. It features a 14-inch diamond blade that can cut concrete, brick, block and stone. The saw has a large cutting capacity of 5-1/8 inches for depth of cut and can make 45-degree miter cuts. It has a dust collection system to help control dust. The saw weighs only 15.7 lbs for easy maneuverability on the job site. The saw's compact size allows it to cut within 2 inches of a wall.
Catalog No. 50-275-0907 Supplier Grainger Supplier No. 14G991
The Makita 14-Inch Masonry Saw has a powerful 15.0 AMP motor that delivers 5,800 RPM for fast, efficient cutting. It features a 14-inch diamond blade that can cut concrete, brick, block and stone. The saw has a large cutting capacity of 5-1/8 inches for depth of cut and can make 45-degree miter cuts. It has a dust collection system to help control dust. The saw weighs only 15.7 lbs for easy maneuverability on the job site. The saw's compact size allows it to cut within 2 inches of a wall.