Learn More
Aviva Systems Biology Corp MASP2 Antibody - N-terminal region (ARP41573_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41573_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MASP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MASP2. Alternate names: MAP19, MASP-2, sMAP, MASP1P1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-57910