Learn More
Aviva Systems Biology Corp Masp2 Antibody - middle region (ARP41575_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41575_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Masp2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: MASP-2, MAp19ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-284-942