Learn More
Medchemexpress LLC HY-P7385 10ug Medchemexpress, Maspin, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP738510UG
Encompass_Preferred
Medchemexpress, HY-P7385 10ug Maspin, Human CAS: Recombinant Human Maspin could inhibit invasion and motility of mammary tumors. Recombinant Human Maspin s also found to be a potent angiogenesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0606