Learn More
Ce Elantech Inc MASPLUS Complete Piston Assembly, Essential Autosampler Component, Includes Seals, 1.31 unit
SDP
Catalog No. NC2585584
Encompass
The MASPLUS Complete Piston is a vital component for Thermo Scientific Autosamplers, ensuring precise sample introduction in analytical workflows. It features an assembly designed for easy replacement and optimal sealing, equipped with necessary seals for reliability. This product enhances the performance of devices like MAS Plus and MAS200R, maintaining integrity and efficiency in elemental analysis. Its compatibility with various Thermo Fisher instruments underscores its critical role in laboratory operations.
Catalog No. NC2585584 Supplier Ce Elantech Inc Supplier No. BRE0032652
The MASPLUS Complete Piston is a vital component for Thermo Scientific Autosamplers, ensuring precise sample introduction in analytical workflows. It features an assembly designed for easy replacement and optimal sealing, equipped with necessary seals for reliability. This product enhances the performance of devices like MAS Plus and MAS200R, maintaining integrity and efficiency in elemental analysis. Its compatibility with various Thermo Fisher instruments underscores its critical role in laboratory operations.