Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Proportional Flowmeter Controller, Mass; 10 L/min Gas
Catalog No. NC1201818
Encompass
Measure 30 standard gases while enjoying versatility and high accuracy from a laminar-based mass flow controller. Accuracy of 0.8% of reading 0.2% full-scale; repeatability of 0.2%. 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM.
Catalog No. NC1201818 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3290771
Measure 30 standard gases while enjoying versatility and high accuracy from a laminar-based mass flow controller. Accuracy of 0.8% of reading 0.2% full-scale; repeatability of 0.2%. 100 to 1 turndown with ranges of 1 SCCM full-scale up to 1000 SLPM.