Learn More
ALICAT SCIENTIFIC MASS FLOW CONTROLLER 5IN, GAS:
Catalog No. NC2097155
Encompass
Mass Flow Controller, 0 - 20 SLPM, Display, Internal Barometer Chip.
This item is not returnable. View return policy
Catalog No. NC2097155 Supplier ALICAT SCIENTIFIC Supplier No. MC20SLPMDIB/5M
Mass Flow Controller, 0 - 20 SLPM, Display, Internal Barometer Chip.