Learn More
Waters Corp Mass Scale and Resolution Set-up Solution
Catalog No. 50201681 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Mass Spectrometry standards kit for system assessment. Mass Scale and Resolution Set Up solution : NaRbI 2.0ug/uL NaI 0.05ug/uL RbI In 50:50 2-Propanol and Water, 15mL Nalgene Screw Top Bottle Certificate of Analysis
Catalog No. 50-201-681 Supplier Waters Corp Supplier No. 700003917
Mass Spectrometry standards kit for system assessment. Mass Scale and Resolution Set Up solution : NaRbI 2.0ug/uL NaI 0.05ug/uL RbI In 50:50 2-Propanol and Water, 15mL Nalgene Screw Top Bottle Certificate of Analysis