Learn More
GOLDEN WEST DIAGNOSTICS Mass Spect Gold Human Urine, Ultra-Low Hormones & Steroids, Toxicology Neg. | 1L
Catalog No. NC1819216
Encompass_Preferred
Source: Human |Country of Origin: USA Origin | Filtration: 0.22 micron | Analyte(s): Reference specification sheet | Analyte Specs: Reference specification sheet | Storage temp: -20 degrees C* | Definition: * Defined as -15C to -30C
Catalog No. NC1819216 Supplier GOLDEN WEST DIAGNOSTICS Supplier No. MSG5000L
Source: Human |Country of Origin: USA Origin | Filtration: 0.22 micron | Analyte(s): Reference specification sheet | Analyte Specs: Reference specification sheet | Storage temp: -20 degrees C* | Definition: * Defined as -15C to -30C