Learn More
IONBENCH CORPORATION Mass Spectrometry Bench 75" x 34.5" (190x88 cm) with integrated Large noise enclosure
Catalog No. 5000150570
Encompass_Preferred
Mass Spectrometry Bench 75" x 34.5" (190x88 cm) with integrated Large noise enclosure. Bundle Includes: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant.DR3 Bank of three drawers. PC1 CPU holder to be fixed on one side of the bench Installable right or left. ARM1 Pack of a single monitor arm and sliding keyboard. ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets. VPSP58 Stainless steel drip pan for vacuum pump To prevent oil spread in case of leakage. SOLV Container for solvent waste bottles.
Catalog No. 50-001-50570 Supplier IONBENCH CORPORATION Supplier No. BCH75INCHNE58
Add to Cart
Add to Cart
Request a Quote
Mass Spectrometry Bench 75" x 34.5" (190x88 cm) with integrated Large noise enclosure. Bundle Includes: TRESPA_WS_MS Countertop upgraded to Trespa Toplab, chemically resistant.DR3 Bank of three drawers. PC1 CPU holder to be fixed on one side of the bench Installable right or left. ARM1 Pack of a single monitor arm and sliding keyboard. ELECUS Power strip, North American 8 outlets fixed behind the bench, NEMA5-15 outlets. VPSP58 Stainless steel drip pan for vacuum pump To prevent oil spread in case of leakage. SOLV Container for solvent waste bottles.