Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REFEYN INC MASSFERENCE P1 CALIBRANT
SDP

Catalog No. NC3245980
Encompass

  • Product type: Protein mass calibrant
  • Calibration range: 90—1000 kDa
  • Instrument compatibility: Refeyn OneMP, TwoMP, TwoMP Auto
  • Purpose: Protein mass calibration
  • Key benefit 1: Accurate measurements
  • Key benefit 2: Reproducible results
  • Format: High concentration aliquots
  • Package contents: 3 x 50 µL aliquots
  • Vial sufficiency: 10 days of calibrations per vial
  • Preparation: Simple to prepare

Catalog No. NC3245980 Supplier REFEYN INC Supplier No. MPCON41033
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Product type: Protein mass calibrant
  • Calibration range: 90—1000 kDa
  • Instrument compatibility: Refeyn OneMP, TwoMP, TwoMP Auto
  • Purpose: Protein mass calibration
  • Key benefit 1: Accurate measurements
  • Key benefit 2: Reproducible results
  • Format: High concentration aliquots
  • Package contents: 3 x 50 µL aliquots
  • Vial sufficiency: 10 days of calibrations per vial
  • Preparation: Simple to prepare

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.