Learn More
REFEYN INC MassFluidix HC - Sample Prep Kit, to support 5 experiments.
SDP
Catalog No. NC3737110
Encompass
"Ready-to-use MassFluidix HC chip to support 5 experiments. This includes:- 1 MassFluidix HC chip (1 chip/pack, 5 measurements/chip) - 2 x bag of mini leur connectors (10/bag)- 1 box of lens Cleaning Tissues (25 sheets)"
Catalog No. NC3737110 Supplier REFEYN INC Supplier No. MPCON21027
"Ready-to-use MassFluidix HC chip to support 5 experiments. This includes:- 1 MassFluidix HC chip (1 chip/pack, 5 measurements/chip) - 2 x bag of mini leur connectors (10/bag)- 1 box of lens Cleaning Tissues (25 sheets)"