Learn More
REFEYN INC MassGlass UC - Auto Sample Prep Kit
SDP
Catalog No. NC3737106
Encompass
"Sample carrier preparation kit to enable 240 sample measurements. Compatible with TwoMP Auto and SamuxMP Auto. This includes:- 10 ready-to-use uncoated and clean sample carrier slides (10 slides/box, 3 months shelf life)- 10 sample well cassettes (24 sample wells/cassette)- 1 box of lens Cleaning Tissues (25 sheets)"
Catalog No. NC3737106 Supplier REFEYN INC Supplier No. MPCON21023
"Sample carrier preparation kit to enable 240 sample measurements. Compatible with TwoMP Auto and SamuxMP Auto. This includes:- 10 ready-to-use uncoated and clean sample carrier slides (10 slides/box, 3 months shelf life)- 10 sample well cassettes (24 sample wells/cassette)- 1 box of lens Cleaning Tissues (25 sheets)"