Learn More
REFEYN INC MassGlass UC - Sample Prep Kit, to enable 60 sample measurements
SDP
Catalog No. NC3737103
Encompass
"Sample carrier preparation kit to enable 60 sample measurements. Compatible with TwoMP, OneMP and SamuxMP. This includes:- 10 ready-to-use uncoated and clean sample carrier slides (10 slides/box, 3 months shelf life)- 10 sample well cassettes (6 sample wells/cassette)- 1 box of lens Cleaning Tissues (25 sheets)"
Catalog No. NC3737103 Supplier REFEYN INC Supplier No. MPCON21022
"Sample carrier preparation kit to enable 60 sample measurements. Compatible with TwoMP, OneMP and SamuxMP. This includes:- 10 ready-to-use uncoated and clean sample carrier slides (10 slides/box, 3 months shelf life)- 10 sample well cassettes (6 sample wells/cassette)- 1 box of lens Cleaning Tissues (25 sheets)"