Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Massoia lactone | 54814-64-1 | 5 G
SDP

Catalog No. 5000421026
Encompass_Preferred

Massoia lactone is a coconut and creamy fragrant compound primarily isolated from *Cryptocarya massoy*. It is also a fragrant biosurfactant produced by the fungus *Aureobasidium pullulans*.

  • Purity: 98.75%
  • Molecular weight: 168.23
  • Molecular formula: C10H16O2
  • Appearance: Liquid
  • Color: Light yellow to light brown
  • Shipping: Room temperature in continental US (may vary elsewhere)
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 200 mg/mL (requires ultrasonic)
  • For research use only

Catalog No. 50-004-21026 Supplier Medchemexpress LLC Supplier No. HYN74355G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Massoia lactone is a coconut and creamy fragrant compound primarily isolated from *Cryptocarya massoy*. It is also a fragrant biosurfactant produced by the fungus *Aureobasidium pullulans*.

  • Purity: 98.75%
  • Molecular weight: 168.23
  • Molecular formula: C10H16O2
  • Appearance: Liquid
  • Color: Light yellow to light brown
  • Shipping: Room temperature in continental US (may vary elsewhere)
  • Storage (pure form): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 200 mg/mL (requires ultrasonic)
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.