Learn More
MilliporeSigma Masson-Goldner Staining Kit
Catalog No. M1004850001 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Reactions:
400 to 500 Stainings
Catalog No. No. of Reactions
M1004850001 400 to 500 Stainings
1 options
This item is not returnable. View return policy
Catalog No. M1004850001 Supplier MilliporeSigma Supplier No. 1.00485.0001
Add to Cart
Add to Cart
This item is not returnable. View return policy
Masson-Goldner staining kit for the visualization of connective tissue with trichromic staining
The Masson-Goldner staining kit for the visualization of connective tissue with trichromic staining, is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin. Using a combination of three different staining solutions, muscle fibers, collagenous fibers, fibrin and erythrocytes can be selectively visualized.
The original methods were primarily used to differentiate collagenous and muscle fibers. The stains used have different molecular sizes and enable the individual tissues to be stained differentially.
The Masson-Goldner staining technique can be carried out using formalin fixed material. Subsequent to staining the nucleus with Weigert's iron hematoxylin, components such as muscle, cytoplasm and erythrocytes are stained with azophloxin and orange G solution. Connective tissue is then counter stained using light green SF solution.
The package is sufficient for 400 to 500 applications. The product is registered as IVD and CE product and can be used in diagnostics and laboratory accreditation.
TRUSTED_SUSTAINABILITY
Specifications
No. of Reactions 400 to 500 Stainings
Product Type Masson-Goldner Stain Kit