Learn More
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit, 500 ml kit
SDP
Catalog No. NC0695533
Encompass
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.
Catalog No. NC0695533 Supplier Newcomer Supply Supplier No. 9179B
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.