Learn More
IMEB, Inc Masson Trichrome Stain Kit, Includes: 1- 8 Oz Bottle/Solutions:Bouin’s, Biecrich’s Scarlet/Acid Fuchsin, Phosphotungstic/Phosphomolybdic Acid, Hematoxylin, 4/2oz Staining Containers, & Storage Tray
SDP
Catalog No. 501689473
Encompass_Preferred
Masson Trichrome stain used for collagen, muscle and pituitary cells. Includes: 1- 8 oz. Bottle Bouin’s solution, 1- 8 oz. Bottle Biecrich’s Scarlet/acid Fuchsin solution, 1- 8 oz. Bottle Phosphotungstic/ Phosphomolybdic acid solution, 1- 8 oz. Bottle Hematoxylin solution, 4- 2 oz. Staining containers and storage tray
Catalog No. 50-168-9473 Supplier IMEB, Inc Supplier No. K7228
Masson Trichrome stain used for collagen, muscle and pituitary cells. Includes: 1- 8 oz. Bottle Bouin’s solution, 1- 8 oz. Bottle Biecrich’s Scarlet/acid Fuchsin solution, 1- 8 oz. Bottle Phosphotungstic/ Phosphomolybdic acid solution, 1- 8 oz. Bottle Hematoxylin solution, 4- 2 oz. Staining containers and storage tray