Learn More
Polysciences, Inc. Masson's Trichrome Stain Kit, 100 or 500 mL
SDP
Catalog No. NC0017745
Encompass
Used for the detection of collagen fibers in tissues such as skin, heart, etc. on formalin-fixed, paraffin-embedded sections and may be used for frozen sections as well. Collagen fibers stained blue, nuclei stained black and cytoplasm, muscle, erythrocytes stained red.
Catalog No. NC0017745 Supplier Polysciences, Inc. Supplier No. 250881
Used for the detection of collagen fibers in tissues such as skin, heart, etc. on formalin-fixed, paraffin-embedded sections and may be used for frozen sections as well. Collagen fibers stained blue, nuclei stained black and cytoplasm, muscle, erythrocytes stained red.