Learn More
Waters Corp Waters Bovine Hemoglobin Digestion Standard, Reversed Phase, UV, MS Detection, Solid
Catalog No. 50781074 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP™ Bovine Hemoglobin digestion standard is prepared by digesting Bovine Hemoglobin (SwissProt HBA P01966, HBB P02081) with sequencing-grade trypsin. This digest is free of undigested protein, trypsin, or other hydrophilic components. Each batch undergoes strict quality control, including a Certificate of Analysis report. This standard is valuable for evaluating HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation.
  • Prepared by digesting bovine hemoglobin with sequencing-grade trypsin.
  • Free of undigested protein, trypsin, and other hydrophilic components.
  • Includes Certificate of Analysis report from strict quality control.
  • Supports reversed phase separation, UV and MS detection.
  • Solid formulation, room temperature storage.
  • Compatible with LC and LCMS systems.
  • Shelf life: 60 months; 1 week upon reconstitution.
Catalog No. 50-781-074 Supplier Waters Corp Supplier No. 186002327
The MassPREP™ Bovine Hemoglobin digestion standard is prepared by digesting Bovine Hemoglobin (SwissProt HBA P01966, HBB P02081) with sequencing-grade trypsin. This digest is free of undigested protein, trypsin, or other hydrophilic components. Each batch undergoes strict quality control, including a Certificate of Analysis report. This standard is valuable for evaluating HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation.
  • Prepared by digesting bovine hemoglobin with sequencing-grade trypsin.
  • Free of undigested protein, trypsin, and other hydrophilic components.
  • Includes Certificate of Analysis report from strict quality control.
  • Supports reversed phase separation, UV and MS detection.
  • Solid formulation, room temperature storage.
  • Compatible with LC and LCMS systems.
  • Shelf life: 60 months; 1 week upon reconstitution.