Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters MassPREP Digestion Standard Kit, Reversed Phase, Solid

Catalog No. 50781077 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The MassPREP™ Protein Digestion Standard Kit contains tryptically digested standards, free of undigested protein, trypsin, or other hydrophilic components. Each batch undergoes strict quality control, with a Certificate of Analysis report. These standards are valuable for evaluating the performance of HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation, verifying LC-MS system performance before separations.

  • Includes five tryptically digested protein standards.
  • Prepared under strict quality control with certificate of analysis.
  • Useful for performance evaluation in HPLC, LC-MS, and mass spectrometry.
  • Separation mode: Reversed phase.
  • Detection method: UV, MS.
  • Formulation: Solid.
  • Shelf life: 60 months.
  • Shelf life upon reconstitution: 1 week.
  • Each standard sufficient for 1000 or more MALDI MS analyses.

Catalog No. 50-781-077 Supplier Waters Corp Supplier No. 186002330
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MassPREP™ Protein Digestion Standard Kit contains tryptically digested standards, free of undigested protein, trypsin, or other hydrophilic components. Each batch undergoes strict quality control, with a Certificate of Analysis report. These standards are valuable for evaluating the performance of HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation, verifying LC-MS system performance before separations.

  • Includes five tryptically digested protein standards.
  • Prepared under strict quality control with certificate of analysis.
  • Useful for performance evaluation in HPLC, LC-MS, and mass spectrometry.
  • Separation mode: Reversed phase.
  • Detection method: UV, MS.
  • Formulation: Solid.
  • Shelf life: 60 months.
  • Shelf life upon reconstitution: 1 week.
  • Each standard sufficient for 1000 or more MALDI MS analyses.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.