Learn More
Waters Corp Waters MassPREP Enhancer, Solid, 5 Vials, Peptide Application
Catalog No. 50782045 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP™ Enhancer is a chemical component designed to enhance phosphopeptide recovery in methods such as those used in phosphopeptide enrichment kits.
  • Solid formulation
  • Suitable for peptide applications
  • Ships under ambient conditions
  • Store at room temperature
  • Provides 36 months of shelf life
  • Each package contains 5 vials
Catalog No. 50-782-045 Supplier Waters Corp Supplier No. 186003863
The MassPREP™ Enhancer is a chemical component designed to enhance phosphopeptide recovery in methods such as those used in phosphopeptide enrichment kits.
  • Solid formulation
  • Suitable for peptide applications
  • Ships under ambient conditions
  • Store at room temperature
  • Provides 36 months of shelf life
  • Each package contains 5 vials