Learn More
Waters Corp Waters MassPREP Digestion Standard, Enolase, Reversed Phase, UV, MS
Catalog No. 50781072 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This digestion standard, from Yeast Enolase with sequencing grade trypsin, provides a protein digest free of undigested components. Strict quality control ensures high quality, with results in a Certificate of Analysis. Ideal for evaluating HPLC, high-performance liquid chromatography, and mass spectrometry instrumentation. Lyophilized powder is room temperature stable; reconstituted solutions stable for 1 week at 8 °C, or extended stability frozen at -20 °C.
  • Reversed phase separation; UV and MS detection.
  • Solid formulation, stable 60 months at room temperature.
  • Reconstituted stable 1 week at 8 °C or longer frozen at -20 °C.
  • Compatible with LC/LCMS for performance and peptide applications.
  • Strict QC with certificate of analysis.
  • Evaluates HPLC columns, MS instrumentation, calibration.
  • Sufficient for 1000+ MALDI MS analyses.
Catalog No. 50-781-072 Supplier Waters Corp Supplier No. 186002325
This digestion standard, from Yeast Enolase with sequencing grade trypsin, provides a protein digest free of undigested components. Strict quality control ensures high quality, with results in a Certificate of Analysis. Ideal for evaluating HPLC, high-performance liquid chromatography, and mass spectrometry instrumentation. Lyophilized powder is room temperature stable; reconstituted solutions stable for 1 week at 8 °C, or extended stability frozen at -20 °C.
  • Reversed phase separation; UV and MS detection.
  • Solid formulation, stable 60 months at room temperature.
  • Reconstituted stable 1 week at 8 °C or longer frozen at -20 °C.
  • Compatible with LC/LCMS for performance and peptide applications.
  • Strict QC with certificate of analysis.
  • Evaluates HPLC columns, MS instrumentation, calibration.
  • Sufficient for 1000+ MALDI MS analyses.