Learn More
Waters Corp Waters Corporation GlycoWorks HILIC μElution Plate, for Glycan Analysis
Catalog No. 50781360 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This plate removes contaminants like salts and detergents from hydrophilic analytes, such as carbohydrates, before mass spectrometry analysis. It enables elution volumes as low as 25 μL, eliminating time-consuming evaporation steps. Eluted carbohydrates can be analyzed directly by MALDI MS without derivatization. It can be used for carbohydrate extraction or as part of a glycan analysis kit to clean glycans released from glycoproteins.
  • Utilizes an optimized HILIC sorbent for high recovery of N-glycans.
  • Suitable for effective labeling reaction clean-up before LC/FLR/MS analysis.
  • Delivers sensitive, robust, and reproducible results.
Catalog No. 50-781-360 Supplier Waters Corp Supplier No. 186002780
This plate removes contaminants like salts and detergents from hydrophilic analytes, such as carbohydrates, before mass spectrometry analysis. It enables elution volumes as low as 25 μL, eliminating time-consuming evaporation steps. Eluted carbohydrates can be analyzed directly by MALDI MS without derivatization. It can be used for carbohydrate extraction or as part of a glycan analysis kit to clean glycans released from glycoproteins.
  • Utilizes an optimized HILIC sorbent for high recovery of N-glycans.
  • Suitable for effective labeling reaction clean-up before LC/FLR/MS analysis.
  • Delivers sensitive, robust, and reproducible results.