Learn More
Waters Corp MassPREP Micro Desalting Column, 2.1 mm Inner Diameter, 5 mm Length, 20 μm Particle Size
Supplier:  Waters Corp 186004032
Encompass_Preferred
The MassPREP on-line desalting column efficiently desalts proteins for LC-MS analyses. It removes non-volatile salts, improving detection sensitivity by preventing ionization suppression. Its reversed-phase phenyl material “traps” proteins, allowing salts to be washed to waste before protein elution into the mass spectrometer.
  • Efficiently desalts proteins for LC-MS analyses.
  • Minimizes non-volatile salts, improving detection sensitivity.
  • Uses reversed-phase phenyl material to separate proteins from salts.
  • Achieves fast cycle times with optimized LC/MS methods.
  • Features 3 μm particles, 100Å pore size, 5 mm length, and 2.1 mm inner diameter.
  • Improves analyte ionization efficiency and sensitivity.
  • Delivers outstanding peak shape and resolution.
  • Designed for hassle-free maintenance.
Catalog No. 50-782-149