Learn More
Waters Corp MassPREP Phenyl Guard Column, 1000Å, 20 µm, 2.1 mm X 10 mm, 10K - 500K, 2/pk
Catalog No. 50781361 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters MassPREP On-Line Desalting Cartridges can effectively desalt proteins prior to LC/MS analyses. Because non-volatile salts (e.g., NaCl) can suppress ionization of intact proteins leading to poor detection sensitivity, it is important to remove or significantly minimize the introduction of these compounds into the mass analyzer. The reversed-phase, phenyl material contained in MassPREP On-Line devices successfully “traps” proteins, allowing the salts to be washed to waste prior to protein elution into the mass spectrometer.
Catalog No. 50-781-361 Supplier Waters Corp Supplier No. 186002785
Waters MassPREP On-Line Desalting Cartridges can effectively desalt proteins prior to LC/MS analyses. Because non-volatile salts (e.g., NaCl) can suppress ionization of intact proteins leading to poor detection sensitivity, it is important to remove or significantly minimize the introduction of these compounds into the mass analyzer. The reversed-phase, phenyl material contained in MassPREP On-Line devices successfully “traps” proteins, allowing the salts to be washed to waste prior to protein elution into the mass spectrometer.