Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Oligonucleotide Standard, 15-35 nt Oligodeoxythymidines, HPLC/UPLC Verification, Lyophilized

Catalog No. 50782176 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This oligonucleotide standard verifies HPLC/UPLC instrument and column performance for synthetic oligonucleotide analysis. It features equimolar amounts of 15, 20, 25, 30, and 35 nt oligodeoxythymidines, lyophilized in 1.5 mL LC vials. Each vial contains approximately 1 nmole of each oligonucleotide. Store at room temperature in its foil pouch.

  • Verifies HPLC/UPLC instrument and column performance.
  • Contains equimolar 15-35 nt oligodeoxythymidines.
  • Lyophilized in 1.5 mL LC vials.
  • Vacuum-sealed to reduce degradation.
  • Approximately 1 nmole of each oligonucleotide per vial.
  • Regularly benchmark column and system performance.
  • 36 months shelf life at room temperature.
  • Solution stable for one week at 4°C after reconstitution.
  • Stability prolonged by storing at -20°C.

Catalog No. 50-782-176 Supplier Waters Corp Supplier No. 186004135
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This oligonucleotide standard verifies HPLC/UPLC instrument and column performance for synthetic oligonucleotide analysis. It features equimolar amounts of 15, 20, 25, 30, and 35 nt oligodeoxythymidines, lyophilized in 1.5 mL LC vials. Each vial contains approximately 1 nmole of each oligonucleotide. Store at room temperature in its foil pouch.

  • Verifies HPLC/UPLC instrument and column performance.
  • Contains equimolar 15-35 nt oligodeoxythymidines.
  • Lyophilized in 1.5 mL LC vials.
  • Vacuum-sealed to reduce degradation.
  • Approximately 1 nmole of each oligonucleotide per vial.
  • Regularly benchmark column and system performance.
  • 36 months shelf life at room temperature.
  • Solution stable for one week at 4°C after reconstitution.
  • Stability prolonged by storing at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.