Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp MassPREP Peptide Mixture, Reversed Phase, UV Detection, Solid, Reference

Catalog No. 50781078 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This peptide standard mixture includes a void volume marker and nine peptides with diverse polarities and isoelectric points. Easily resolved under reversed-phase HPLC conditions, it's ideal for evaluating HPLC columns and systems.

  • Contains a void volume marker and nine peptides.
  • Wide range of polarities and isoelectric points.
  • Easily resolved under reversed-phase HPLC conditions.
  • Useful for evaluating HPLC columns and systems.
  • Validates system setup and improves methodology confidence.
  • Streamlines workflow, reduces sample prep time.
  • Assesses column and system performance.
  • Separation mode: Reversed Phase.
  • Detection method: UV.
  • Formulation: Solid.
  • Storage: Room temperature.
  • System type: LC.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping: Ambient.
  • Type: Reference.

Catalog No. 50-781-078 Supplier Waters Corp Supplier No. 186002337
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This peptide standard mixture includes a void volume marker and nine peptides with diverse polarities and isoelectric points. Easily resolved under reversed-phase HPLC conditions, it's ideal for evaluating HPLC columns and systems.

  • Contains a void volume marker and nine peptides.
  • Wide range of polarities and isoelectric points.
  • Easily resolved under reversed-phase HPLC conditions.
  • Useful for evaluating HPLC columns and systems.
  • Validates system setup and improves methodology confidence.
  • Streamlines workflow, reduces sample prep time.
  • Assesses column and system performance.
  • Separation mode: Reversed Phase.
  • Detection method: UV.
  • Formulation: Solid.
  • Storage: Room temperature.
  • System type: LC.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping: Ambient.
  • Type: Reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.