Learn More
Waters Corp MassPREP Peptide Mixture, 5/pk
Supplier:  Waters Corp 186002338
Encompass_Preferred
The MassPREP peptide standard mixture 5 pack contains 5 pieces of 186002337. Each standard contains a void volume marker and nine, carefully selected peptides with a broad range of polarities and isoelectric points. All peaks are easily resolved under reversed-phase HPLC conditions typically used for peptide separations. The MassPREP Peptide Standard Mixture is useful for the evaluation of HPLC columns and systems dedicated for peptide separations.
Catalog No. 50-781-079