Learn More
Waters Corp Waters MassPREP Phosphopep Enrich Elute Plate, 96-Well, Affinity Sorbent
Catalog No. 50782037 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This application kit is designed to streamline the isolation and enrichment of phosphopeptides from complex biological samples. This 96-well plate utilizes a specialized phosphopeptide affinity sorbent and includes a unique chemical enhancer to improve the selectivity of solid-phase extraction (SPE). It is ideally suited for preparing low-abundance phosphopeptides for subsequent mass spectrometry (MS) analysis, offering enhanced performance and ease of use.
  • Streamlined isolation of phosphopeptides
  • 96-well format for high-throughput applications
  • Utilizes a specialized phosphopeptide affinity sorbent
  • Includes a chemical enhancer for improved selectivity
  • Enhances solid-phase extraction (SPE) efficiency
  • Prepares low-abundance phosphopeptides for mass spectrometry analysis
  • Offers high selectivity and ease of use
Catalog No. 50-782-037 Supplier Waters Corp Supplier No. 186003820
This application kit is designed to streamline the isolation and enrichment of phosphopeptides from complex biological samples. This 96-well plate utilizes a specialized phosphopeptide affinity sorbent and includes a unique chemical enhancer to improve the selectivity of solid-phase extraction (SPE). It is ideally suited for preparing low-abundance phosphopeptides for subsequent mass spectrometry (MS) analysis, offering enhanced performance and ease of use.
  • Streamlined isolation of phosphopeptides
  • 96-well format for high-throughput applications
  • Utilizes a specialized phosphopeptide affinity sorbent
  • Includes a chemical enhancer for improved selectivity
  • Enhances solid-phase extraction (SPE) efficiency
  • Prepares low-abundance phosphopeptides for mass spectrometry analysis
  • Offers high selectivity and ease of use