Learn More
Waters Corp WATERS CORPORATION Phosphopeptide Enrichment Kit, 96-well, SPE
Catalog No. 50782046 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP™ Phosphopeptide Enrichment Kit is designed for the selective enrichment of phosphopeptides from complex samples. It includes a 96-well micro-scale solid-phase extraction (SPE) plate packed with an affinity sorbent and a unique chemical (Enhancer™) to improve selectivity. This method simplifies phosphopeptide enrichment by not requiring pre-loading the sorbent with a metal chelator, unlike immobilized metal affinity chromatography (IMAC) technology.
  • Includes a 96-well micro-scale solid-phase extraction (SPE) plate.
  • Uses an affinity sorbent.
  • Includes Enhancer™ for improved selectivity.
  • Does not require pre-loading the sorbent with metal chelator.
Catalog No. 50-782-046 Supplier Waters Corp Supplier No. 186003864
The MassPREP™ Phosphopeptide Enrichment Kit is designed for the selective enrichment of phosphopeptides from complex samples. It includes a 96-well micro-scale solid-phase extraction (SPE) plate packed with an affinity sorbent and a unique chemical (Enhancer™) to improve selectivity. This method simplifies phosphopeptide enrichment by not requiring pre-loading the sorbent with a metal chelator, unlike immobilized metal affinity chromatography (IMAC) technology.
  • Includes a 96-well micro-scale solid-phase extraction (SPE) plate.
  • Uses an affinity sorbent.
  • Includes Enhancer™ for improved selectivity.
  • Does not require pre-loading the sorbent with metal chelator.