Learn More
Waters Corp Waters Phosphopeptide Standard, Enolase, Phosphorylated Peptides, Reference
Catalog No. 50781652 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP™ Phosphopeptide Standard Enolase is a mix of four Yeast Enolase-derived phosphorylated peptides (three singly, one doubly phosphorylated). This standard optimizes phosphopeptide detection in LC/MS, LC/UV, and MALDI-MS, allowing control over sample preparation and peptide ratios.
  • Optimizes workflows for phosphopeptide detection.
  • Verifies system performance on liquid chromatography and mass spectrometry systems.
  • Solid formulation.
  • Store at room temperature.
  • Shelf life of 24 months; 1 week upon reconstitution.
  • Ships ambient.
  • Reference standard.
  • Reversed phase separation mode.
  • UV and MS detection methods.
  • Compatible with LC and LCMS systems.
  • Includes test results for purity and quality control.
  • One unit per package.
Catalog No. 50-781-652 Supplier Waters Corp Supplier No. 186003285
The MassPREP™ Phosphopeptide Standard Enolase is a mix of four Yeast Enolase-derived phosphorylated peptides (three singly, one doubly phosphorylated). This standard optimizes phosphopeptide detection in LC/MS, LC/UV, and MALDI-MS, allowing control over sample preparation and peptide ratios.
  • Optimizes workflows for phosphopeptide detection.
  • Verifies system performance on liquid chromatography and mass spectrometry systems.
  • Solid formulation.
  • Store at room temperature.
  • Shelf life of 24 months; 1 week upon reconstitution.
  • Ships ambient.
  • Reference standard.
  • Reversed phase separation mode.
  • UV and MS detection methods.
  • Compatible with LC and LCMS systems.
  • Includes test results for purity and quality control.
  • One unit per package.