Learn More
Waters Corp Waters MassPREP Phosphorylase b Standard, Solid, Reversed Phase, UV, MS
Catalog No. 50781073 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP™ Phosphorylase b digestion standard from Waters is prepared by digesting Phosphorylase b (SwissProt P00489) with sequencing grade trypsin. This standard is useful for evaluating HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation.
  • Solid formulation.
  • Store at room temperature.
  • Shelf life of 60 months; 1 week upon reconstitution.
  • Reversed phase separation mode.
  • UV and MS detection methods.
  • Compatible with LC and LCMS systems.
  • Digestion standard for system performance applications.
  • Prepared from Phosphorylase b (SwissProt P00489) with sequencing grade trypsin.
  • Protein digest free of undigested standard protein, trypsin, or other hydrophilic components.
  • Includes Certificate of Analysis report.
Catalog No. 50-781-073 Supplier Waters Corp Supplier No. 186002326
The MassPREP™ Phosphorylase b digestion standard from Waters is prepared by digesting Phosphorylase b (SwissProt P00489) with sequencing grade trypsin. This standard is useful for evaluating HPLC columns, high-performance liquid chromatography, and mass spectrometry instrumentation.
  • Solid formulation.
  • Store at room temperature.
  • Shelf life of 60 months; 1 week upon reconstitution.
  • Reversed phase separation mode.
  • UV and MS detection methods.
  • Compatible with LC and LCMS systems.
  • Digestion standard for system performance applications.
  • Prepared from Phosphorylase b (SwissProt P00489) with sequencing grade trypsin.
  • Protein digest free of undigested standard protein, trypsin, or other hydrophilic components.
  • Includes Certificate of Analysis report.