Learn More
Waters Corp Waters MassPREP Digestion Standard Mix 2, Tryptically Digested Proteins, 60 Months Shelf Life
Catalog No. 50781409 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The MassPREP Digestion Standard Mix 2 provides molar ratios for tryptically digested proteins: Yeast Enolase, Phosphorylase b, Yeast Alcohol Dehydrogenase (ADH), and Bovine Serum Albumin (BSA). The precise ratio is on the Certificate of Analysis (COA). This standard verifies PLGS software and assesses HPLC, LC, and mass spectrometry equipment performance.
  • Includes tryptically digested proteins: Yeast Enolase, Phosphorylase b, Yeast Alcohol Dehydrogenase (ADH), Bovine Serum Albumin (BSA).
  • Exact molar ratio available on Certificate of Analysis (COA).
  • Quality-control approved peptide standard.
  • Shelf life up to 60 months or one week after reconstitution.
  • Store at room temperature.
  • Verifies PLGS software.
  • Assesses HPLC, LC, mass spectrometry equipment performance.
  • Evaluates system performance for valuable materials.
Catalog No. 50-781-409 Supplier Waters Corp Supplier No. 186002866
The MassPREP Digestion Standard Mix 2 provides molar ratios for tryptically digested proteins: Yeast Enolase, Phosphorylase b, Yeast Alcohol Dehydrogenase (ADH), and Bovine Serum Albumin (BSA). The precise ratio is on the Certificate of Analysis (COA). This standard verifies PLGS software and assesses HPLC, LC, and mass spectrometry equipment performance.
  • Includes tryptically digested proteins: Yeast Enolase, Phosphorylase b, Yeast Alcohol Dehydrogenase (ADH), Bovine Serum Albumin (BSA).
  • Exact molar ratio available on Certificate of Analysis (COA).
  • Quality-control approved peptide standard.
  • Shelf life up to 60 months or one week after reconstitution.
  • Store at room temperature.
  • Verifies PLGS software.
  • Assesses HPLC, LC, mass spectrometry equipment performance.
  • Evaluates system performance for valuable materials.